Diethyl maleate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Maleic acid diethyl ester, Diethyl (Z)-2-butenedioate
L03876-06
L03876-0B
L03876-30
AAAL03876-06EA
24.7
USD
AAAL03876-06
AAAL03876-0B
AAAL03876-30
Diethyl maleate 97%
Diethyl maleate
Purity: 97%
CAS No.: 141-05-9
Molecular Formula: C8H12O4
Formula Weight: 172.18
Flash Point: 93°(199°F)
Refractive Index: 1.4400
Odor: Pleasant
Melting Point: -11°
Boiling Point: 221-224°
Beilstein Reference: 1100825
Merck Reference: 14,3123
MDL No.: MFCD00009191
Formula:
C₈H₁₂O₄ MW: 172.18 g/mol Boiling Pt: 225 Melting Pt: -11 Density: 1.064 Flash Pt: 93 °C(199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009191 CAS Number: 141-05-9 EINECS: 205-451-9 Merck Index: 14,3123 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...