tert-Pentylbenzene 97%
Supplier: Thermo Scientific Chemicals
Warning
|
L03731-09
L03731-18
AAAL03731-09EA
49.1
USD
AAAL03731-09
AAAL03731-18
tert-Pentylbenzene 97%
tert-Pentylbenzene
Purity: 97%
CAS No.: 2049-95-8
Molecular Formula: C11H16
Formula Weight: 148.25
Flash Point: 65°(149°F)
Refractive Index: 1.4950
Boiling Point: 188-191°
Beilstein Reference: 2039053
MDL No.: MFCD00015187
Formula:
C₆H₅C(CH₃)₂CH₂CH₃ MW: 148.25 g/mol Boiling Pt: 188…191 °C Density: 0.8889 g/cm³ (0 °C) Flash Pt: 65 °C(149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015187 CAS Number: 2049-95-8 EINECS: 218-076-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...