α-Methylstyrene 99% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Phenylpropene, Isopropenylbenzene
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC) 10-20 ppm
L03609-0F
L03609-AE
L03609-AP
AAAL03609-0FEA
119
USD
AAAL03609-0F
AAAL03609-AE
AAAL03609-AP
α-Methylstyrene 99% stabilized
α-Methylstyrene
Purity: 99%
CAS No.: 98-83-9
Molecular Formula: C9H10
Formula Weight: 118.18
Flash Point: 45°(113°F)
Refractive Index: 1.5380
Melting Point: -23°
Boiling Point: 166-169°
Beilstein Reference: 969405
UN No.: 2303
MDL No.: MFCD00008859
Formula:
C₉H₁₀ MW: 118.18 g/mol Boiling Pt: 166…169 °C Melting Pt: –23 °C Density: 0.9108 g/cm³ (20 °C) Flash Pt: 45 °C (113 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008859 CAS Number: 98-83-9 EINECS: 202-705-0 UN: 2303 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...