1,4-Butanediol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03491-0B
L03491-30
L03491-0E
AAL03491-0BEA
42.26
USD
AAL03491-0B
AAL03491-30
AAL03491-0E
1,4-Butanediol 99%
1,4-Butanediol
Purity: 99%
CAS No.: 110-63-4
Molecular Formula: HO(CH2)4OH
Formula Weight: 90.12
Flash Point: 135°(275°F)
Refractive Index: 1.4460
Melting Point: 19-20°
Boiling Point: 229-230°
Beilstein Reference: 1633445
MDL No.: MFCD00002968
Formula:
C₄H₁₀O₂ Boiling Pt: 229…230 °C Melting Pt: 19…20 °C Density: 1.017 Flash Pt: 135 °C(275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002968 CAS Number: 110-63-4 EINECS: 203-786-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...