1-Methylpiperidine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Methylpiperidine
L03398-22
L03398-09
L03398-36
AAAL03398-22EA
50.6
USD
AAAL03398-22
AAAL03398-09
AAAL03398-36
1-Methylpiperidine 99%
1-Methylpiperidine
Purity: 99%
CAS No.: 626-67-5
Molecular Formula: C6H13N
Formula Weight: 99.18
Flash Point: 3°(37°F)
Refractive Index: 1.4378
Odor: Amine-like
pH: 12
Melting Point: -50°
Boiling Point: 105-107°
Beilstein Reference: 1073
UN No.: 2399
MDL No.: MFCD00006491
Formula:
C₆H₁₃N MW: 99.18 g/mol Boiling Pt: 105…107 °C Melting Pt: –50 °C Density: 0.82 g/cm³ (20 °C) Flash Pt: 3 °C (37 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006491 CAS Number: 626-67-5 EINECS: 210-959-9 UN: 2399 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...