Tetraethylsilane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Purity: 97%
CAS No.: 631-36-7
Molecular Formula: C8H20Si
Formula Weight: 144.33
Flash Point: 26°(78°F)
Refractive Index: 1.4260
Odor: Recognizable
Melting Point: -82°
Boiling Point: 153-155°
Beilstein Reference: 1732768
UN No.: 1993
MDL No.: MFCD00009019
Formula:
C₈H₂₀Si Boiling Pt: 153…155 °C Melting Pt: –82 °C Density: 0.767 Flash Pt: 26 °C (78 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009019 CAS Number: 631-36-7 EINECS: 211-155-0 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...