4-Bromo-3-chloroaniline 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03268-18
L03268-09
AAAL03268-18EA
251
USD
AAAL03268-18
AAAL03268-09
4-Bromo-3-chloroaniline 96%
4-Bromo-3-chloroaniline
Purity: 96%
CAS No.: 21402-26-6
Molecular Formula: C6H5BrClN
Formula Weight: 206.47
Odor: Characteristic
Melting Point: 61-66°
Beilstein Reference: 1210160
UN No.: 2811
MDL No.: MFCD00041311
Formula:
C₆H₅BrClN Boiling Pt: 264 °C (1013 hPa) Melting Pt: 61…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041311 CAS Number: 21402-26-6 EINECS: 244-370-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...