4-Cyclohexylaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

L03184-03 L03184-06
AAAL03184-03EA 64.4 USD
AAAL03184-03 AAAL03184-06
4-Cyclohexylaniline 97%
4-Cyclohexylaniline

Purity: 97% CAS No.: 6373-50-8 Molecular Formula: C12H17N Formula Weight: 175.28 Melting Point: 52-56° Boiling Point: 166°/13mm MDL No.: MFCD00001454

Formula: C₁₂H₁₇N
MW: 175.27 g/mol
Boiling Pt: 166 °C/13 mm
Melting Pt: 52…56 °C
Density: 1.13 g/cm³ (-100.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001454
CAS Number: 6373-50-8
EINECS: 228-918-9
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR