Tetrabutoxysilane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Purity: 97%
CAS No.: 4766-57-8
Molecular Formula: C16H36O4Si
Formula Weight: 320.55
Flash Point: 135°(275°F)
Refractive Index: 1.4130
Odor: Characteristic
Melting Point:
Formula:
C₁₆H₃₆O₄Si Boiling Pt: 277 °C Melting Pt: < –80 °C Density: 0.9353 g/cm³ (0 °C) Flash Pt: 135 °C(275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009432 CAS Number: 4766-57-8 EINECS: 225-305-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...