3-Methylpentane 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 99+%
CAS No.: 96-14-0
Molecular Formula: C6H14
Formula Weight: 86.18
Flash Point: -32°(-26°F)
Refractive Index: 1.3770
Odor: Petroleum-like
Melting Point: -118°
Boiling Point: 63-65°
Beilstein Reference: 1730734
UN No.: 1208
MDL No.: MFCD00009342
Formula:
C₆H₁₄ Boiling Pt: 63…65 °C Melting Pt: –118 °C Density: 0.663 Flash Pt: -32 °C(-26 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009342 CAS Number: 96-14-0 EINECS: 202-481-4 UN: 1208 ADR: "3",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...