3-Phenylpropylamine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Phenyl-1-propylamine
L03023-09
L03023-18
AAAL03023-09EA
43.2
USD
AAAL03023-09
AAAL03023-18
3-Phenylpropylamine 98+%
3-Phenylpropylamine
Purity: 98+%
CAS No.: 2038-57-5
Molecular Formula: C6H5CH2CH2CH2NH2
Formula Weight: 135.21
Flash Point: 90°(194°F)
Refractive Index: 1.5250
Odor: Amine-like
Boiling Point: 220-221°
Beilstein Reference: 2205526
UN No.: 2735
MDL No.: MFCD00008224
Formula:
C₉H₁₃N MW: 135.21 g/mol Boiling Pt: 221 °C (1013 hPa) Density: 0.951 g/cm³ (15 °C) Flash Pt: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008224 CAS Number: 2038-57-5 EINECS: 218-012-1 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...