2-Methylpentane 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,1-Dimethylbutane, Isohexane
L03001-AE
L03001-AP
AAAL03001-AEEA
81.33
USD
AAAL03001-AE
AAAL03001-AP
2-Methylpentane 99+%
2-Methylpentane
Purity: 99+%
CAS No.: 107-83-5
Molecular Formula: C6H14
Formula Weight: 86.18
Flash Point: -40°(-40°F)
Refractive Index: 1.3710
Odor: Petroleum-like
Melting Point: -154°
Boiling Point: 61-62°
Beilstein Reference: 1730735
UN No.: 1208
MDL No.: MFCD00009406
Formula:
C₆H₁₄ MW: 86.18 g/mol Boiling Pt: 61…62 °C Melting Pt: –154 °C Density: 0.653 g/cm³ (20 °C) Flash Pt: –40 °C (–40 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009406 CAS Number: 107-83-5 EINECS: 203-523-4 UN: 1208 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...