3-Pentyn-1-ol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pent-3-yn-1-ol

L02951-03 L02951-06
AAL02951-03EA 32.5 USD
AAL02951-03 AAL02951-06
3-Pentyn-1-ol 98%
3-Pentyn-1-ol

Purity: 98% CAS No.: 10229-10-4 Molecular Formula: C5H8O Formula Weight: 84.12 Flash Point: 54°(129°F) Refractive Index: 1.4560 Odor: Recognizable Boiling Point: 154-157° Beilstein Reference: 1735967 UN No.: 1987 MDL No.: MFCD00002956

Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 154…157 °C
Density: 0.9002 g/cm³ (20 °C)
Flash Pt: 54 °C (129 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002956
CAS Number: 10229-10-4
EINECS: 233-550-7
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR