1-Undecyne 97%
Supplier: Thermo Scientific Chemicals
Warning
|
L02883-18
L02883-09
AAAL02883-18EA
462
USD
AAAL02883-18
AAAL02883-09
1-Undecyne 97%
1-Undecyne
Purity: 97%
CAS No.: 2243-98-3
Molecular Formula: C11H20
Formula Weight: 152.28
Flash Point: 65°(149°F)
Refractive Index: 1.4310
Odor: Characteristic
Melting Point: -25°
Boiling Point: 196°
Beilstein Reference: 1740043
MDL No.: MFCD00015063
Formula:
C₁₁H₂₀ MW: 152.28 g/mol Boiling Pt: 196 °C Melting Pt: –25 °C Density: 0.7728 g/cm³ (20 °C) Flash Pt: 65 °C(149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015063 CAS Number: 2243-98-3 EINECS: 218-825-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...