2,3,5-Triiodobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: TIBA

L02679-06 L02679-22 L02679-14
AAAL02679-06EA 51.2 USD
AAAL02679-06 AAAL02679-22 AAAL02679-14
2,3,5-Triiodobenzoic acid 98+%
2,3,5-Triiodobenzoic acid

Purity: 98+% CAS No.: 88-82-4 Molecular Formula: C7H3I3O2 Formula Weight: 499.81 Melting Point: 220-222° Beilstein Reference: 1955088

Formula: C₇H₃I₃O₂
MW: 499.81 g/mol
Boiling Pt: 416 °C (1013 hPa)
Melting Pt: 220…222 °C
Storage Temperature: Freezer
MDL Number: MFCD00002420
CAS Number: 88-82-4
EINECS: 201-859-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR