tert-Butyl isothiocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

L02668-06 L02668-14
AAAL02668-06EA 59.4 USD
AAAL02668-06 AAAL02668-14
tert-Butyl isothiocyanate 97%
tert-Butyl isothiocyanate

Purity: 97% CAS No.: 590-42-1 Molecular Formula: (CH3)3CNCS Formula Weight: 115.20 Flash Point: 38°(100°F) Refractive Index: 1.4810 Odor: Pungent Melting Point: 10-12° Boiling Point: 141-145° Beilstein Reference: 1361854 UN No.: 2929 MDL No.: MFCD00004816

Formula: C₅H₉NS
MW: 115.2 g/mol
Boiling Pt: 141…145 °C
Melting Pt: 10…12 °C
Density: 0.908
Flash Pt: 38 °C (100 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004816
CAS Number: 590-42-1
EINECS: 209-682-6
UN: 2929
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR