2-Hexynoic acid 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02526-03
L02526-06
AAAL02526-03EA
25.69
USD
AAAL02526-03
AAAL02526-06
2-Hexynoic acid 96%
2-Hexynoic acid
Purity: 96%
CAS No.: 764-33-0
Molecular Formula: CH3(CH2)2C≡CCO2H
Formula Weight: 112.13
Refractive Index: 1.4590
Melting Point: 23-26°
Boiling Point: 110-112°/10mm
Beilstein Reference: 1744983
UN No.: 3261
MDL No.: MFCD00014371
Formula:
C₆H₈O₂ Boiling Pt: 110…112 °C/10 mm Melting Pt: 23…26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014371 CAS Number: 764-33-0 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...