5-Aminoindan 98+%

Supplier: Thermo Scientific Chemicals
Warning

L02504-18 L02504-09
AAAL02504-18EA 375 USD
AAAL02504-18 AAAL02504-09
5-Aminoindan 98+%
5-Aminoindan

Purity: 98+% CAS No.: 24425-40-9 Molecular Formula: C9H11N Formula Weight: 133.19 Flash Point: 110°(230°F) Melting Point: 32-37° Boiling Point: 248-250° Beilstein Reference: 2082278 UN No.: 2811 MDL No.: MFCD00003803

Formula: C₉H₁₁N
Boiling Pt: 248…250 °C
Melting Pt: 32…37 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003803
CAS Number: 24425-40-9
EINECS: 246-241-7
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR