Fluorene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
9H-Fluorene, Diphenylenmethane
L02124-22
L02124-36
L02124-14
AAAL02124-22EA
80.73
USD
AAAL02124-22
AAAL02124-36
AAAL02124-14
Fluorene 98+%
Fluorene
Purity: 98+%
CAS No.: 86-73-7
Molecular Formula: C13H10
Formula Weight: 166.22
Flash Point: 151°(303°F)
Melting Point: 113-117°
Boiling Point: 295°
Beilstein Reference: 1363491
Merck Reference: 14,4155
UN No.: 3077
MDL No.: MFCD00001111
Formula:
C₁₃H₁₀ MW: 166.22 g/mol Boiling Pt: 295 °C Melting Pt: 113…117 °C Density: 1.203 g/cm³ (20 °C) Flash Pt: 151 °C (303 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001111 CAS Number: 86-73-7 EINECS: 201-695-5 UN: 3077 ADR: 9,III Merck Index: 13,04181 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...