2',3',4',5',6'-Pentafluoroacetophenone 97%
Supplier: Thermo Scientific Chemicals
Warning
|
L02063-06
L02063-03
AAAL02063-06EA
86.1
USD
AAAL02063-06
AAAL02063-03
2',3',4',5',6'-Pentafluoroacetophenone 97%
2',3',4',5',6'-Pentafluoroacetophenone
Purity: 97%
CAS No.: 652-29-9
Molecular Formula: C8H3F5O
Formula Weight: 210.10
Flash Point: 65°(149°F)
Refractive Index: 1.4366
Boiling Point: 130-131°
Beilstein Reference: 2053225
MDL No.: MFCD00000296
Formula:
C₈H₃F₅O MW: 210.1 g/mol Boiling Pt: 130…131 °C Density: 1.47 g/cm³ (20 °C) Flash Pt: 65 °C(149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000296 CAS Number: 652-29-9 EINECS: 211-487-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...