2-Ethylphenol 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Ethyl-2-hydroxybenzene
L01637-18
L01637-09
AAAL01637-18EA
102
USD
AAAL01637-18
AAAL01637-09
2-Ethylphenol 98+%
2-Ethylphenol
Purity: 98+%
CAS No.: 90-00-6
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 78°(172°F)
Refractive Index: 1.5360
Melting Point: -18°
Boiling Point: 206°
Beilstein Reference: 1099397
Merck Reference: 14,3839
UN No.: 3145
MDL No.: MFCD00002249
Formula:
C₈H₁₀O MW: 122.17 g/mol Boiling Pt: 204.5 °C (1013 hPa) Melting Pt: -18 °C Density: 1.0371 g/cm³ (0 °C) Flash Pt: 78 °C (172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002249 CAS Number: 90-00-6 EINECS: 201-958-4 UN: 3145 ADR: 8,III Merck Index: 13,03871 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...