1-Ethoxy-2,2,2-trifluoroethanol tech. 80%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Trifluoroacetaldehyde ethyl hemiacetal, Trifluoroacetaldehydeethylhemiacetal
L01554-06
L01554-14
AAAL01554-06EA
70.4
USD
AAAL01554-06
AAAL01554-14
1-Ethoxy-2,2,2-trifluoroethanol tech. 80%, Technical Grade
1-Ethoxy-2,2,2-trifluoroethanol
Purity: 80%
CAS No.: 433-27-2
Molecular Formula: C4H7F3O2
Formula Weight: 144.09
Flash Point: 39°(102°F)
Refractive Index: 1.3430
Odor: Odorless
Boiling Point: 104-106°
Beilstein Reference: 906797
UN No.: 1989
MDL No.: MFCD00000441
Formula:
C₄H₇F₃O₂ MW: 144.09 g/mol Boiling Pt: 104…106 °C Density: 1.230 Flash Pt: 39 °C(102 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000441 CAS Number: 433-27-2 EINECS: 207-087-6 UN: 1989 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...