4-Methylstyrene 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
p-Vinyltoluene, p-Methylstyrene, 4-Vinyltoluene
Stabilization:
Stabilised with 3,5-di-tert-butylpyrocatechol 0.1%
L01480-09
L01480-18
AAAL01480-09EA
36.24
USD
AAAL01480-09
AAAL01480-18
4-Methylstyrene 98% stabilized
4-Methylstyrene
Purity: 98%
CAS No.: 622-97-9
Molecular Formula: C9H10
Formula Weight: 118.18
Storage Temperature: 2°C to 8°C
Flash Point: 45°(113°F)
Refractive Index: 1.5430
Melting Point: -34°
Boiling Point: 171-175°
Beilstein Reference: 1209317
UN No.: 2618
MDL No.: MFCD00008621
Formula:
C₉H₁₀ MW: 118.18 g/mol Boiling Pt: 171…175 °C Melting Pt: –34 °C Density: 0.9383 g/cm³ (0 °C) Flash Pt: 45 °C (113 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008621 CAS Number: 622-97-9 EINECS: 210-762-8 UN: 2618 ADR: 3,III |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...