2-Phenylpropionic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (±)-2-Phenylpropionic acid, (±)-Hydratropic acid, DL-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid

L00709-14 L00709-06 L00709-22
AAAL00709-14EA 124 USD
AAAL00709-14 AAAL00709-06 AAAL00709-22
2-Phenylpropionic acid 98%
2-Phenylpropionic acid

Purity: 98% CAS No.: 492-37-5 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 148°(298°F) Refractive Index: 1.5230 Melting Point: -5° Boiling Point: 264-266° Beilstein Reference: 1863558 MDL No.: MFCD00002650

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 264…266 °C
Melting Pt: –5 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 148 °C(298 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002650
CAS Number: 492-37-5
EINECS: 207-752-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR