Tetrabromopyrocatechol 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetrabromocatechol, Tetrabromobenzene-1,2-diol

L00690-14 L00690-06
AAAL00690-14EA 103 USD
AAAL00690-14 AAAL00690-06
Tetrabromopyrocatechol 96%
Tetrabromopyrocatechol

Purity: 96% CAS No.: 488-47-1 Molecular Formula: C6H2Br4O2 Formula Weight: 425.70 Melting Point: 189-193° Beilstein Reference: 2051548

Formula: C₆H₂Br₄O₂
MW: 425.7 g/mol
Boiling Pt: 379 °C (1013 hPa)
Melting Pt: 189…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00002189
CAS Number: 488-47-1
EINECS: 207-678-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR