N,N-Dimethylformamide diethyl acetal 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,1-Diethoxy-N,N-dimethylmethanamine, 1,1-Diethoxytrimethylamine
L00205-09
L00205-18
AAAL00205-09EA
32.8
USD
AAAL00205-09
AAL00205-18
N,N-Dimethylformamide diethyl acetal 95%
N,N-Dimethylformamide diethyl acetal
Purity: 95%
CAS No.: 1188-33-6
Molecular Formula: (CH3)2NCH(OCH2CH3)2
Formula Weight: 147.22
Flash Point: 17°(62°F)
Refractive Index: 1.3990
Boiling Point: 134-136°
Beilstein Reference: 741889
UN No.: 1993
MDL No.: MFCD00009227
Formula:
C₇H₁₇NO₂ Boiling Pt: 134…136 °C Density: 0.86 g/cm³ (20 °C) Flash Pt: 17 °C(62 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009227 CAS Number: 1188-33-6 EINECS: 214-707-9 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...