2-Chloro-1-methylimidazole 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H51867-03
H51867-MD
AAH51867-03EA
187
USD
AAH51867-03
AAH51867-MD
2-Chloro-1-methylimidazole 97+%
2-Chloro-1-methylimidazole
Purity: 97+%
CAS No.: 253453-91-7
Molecular Formula: C4H5ClN2
Formula Weight: 116.55
Melting Point: -10°
MDL No.: MFCD02179531
Formula:
C₄H₅ClN₂ Melting Pt: –10 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179531 CAS Number: 253453-91-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...