2-Chloroimidazole 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Chloro-1H-imidazole

H51863-MD H51863-03
AAH51863-MDEA 92.5 USD
AAH51863-MD AAH51863-03 AAH51863-06
2-Chloroimidazole 97%
2-Chloroimidazole

Purity: 97% CAS No.: 16265-04-6 Molecular Formula: C3H3ClN2 Formula Weight: 102.52 Melting Point: 165-166° MDL No.: MFCD02179530

Formula: C₃H₃ClN₂
MW: 102.52
Melting Pt: 165…166 °C
Storage Temperature: Ambient
MDL Number: MFCD02179530
CAS Number: 16265-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR