2-Iodo-1-methylimidazole 97%

Supplier: Thermo Scientific Chemicals
Warning

H51862-MD H51862-03
AAH51862-MDEA 57.6 USD
AAH51862-MD AAH51862-03
2-Iodo-1-methylimidazole 97%
2-Iodo-1-methylimidazole

Purity: 97% CAS No.: 37067-95-1 Molecular Formula: C4H5IN2 Formula Weight: 208.00 Melting Point: 79-82° MDL No.: MFCD00661606

Formula: C₄H₅IN₂
Melting Pt: 79…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00661606
CAS Number: 37067-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR