4-Aminophenylboronic acid pinacol ester 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Aminobenzeneboronic acid pinacol ester

H51666-03 H51666-06
AAH51666-03EA 70.4 USD
AAH51666-03 AAH51666-06
4-Aminophenylboronic acid pinacol ester 98%
4-Aminophenylboronic acid pinacol ester

Purity: 98% CAS No.: 214360-73-3 Molecular Formula: C12H18BNO2 Formula Weight: 219.09 MDL No.: MFCD02093721

Formula: C₁₂H₁₈BNO₂
MW: 219.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093721
CAS Number: 214360-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR