4-Iodophenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(4-Iodophenyl)boronic acid, Dihydroxy(4-iodophenyl)borane, 4-Iodobenzeneboronic acid, (4-Iodophenyl)metaboric acid
H51119-03
H51119-06
H51119-14
AAH51119-03EA
29.6
USD
AAH51119-03
AAH51119-06
AAH51119-14
4-Iodophenylboronic acid 97%
4-Iodophenylboronic acid
Purity: 97%
CAS No.: 5122-99-6
Molecular Formula: C6H6BIO2
Formula Weight: 247.83
MDL No.: MFCD02093073
Formula:
C₆H₆BIO₂ Storage Temperature: Ambient |
MDL Number:
MFCD01319014 CAS Number: 5122-99-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...