3-Iodophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dihydroxy(3-iodophenyl)borane, (3-Iodophenyl)boronic acid, 3-Iodobenzeneboronic acid, (3-Iodophenyl)metaboric acid

H51112-03 H51112-06
AAH51112-03EA 27.6 USD
AAH51112-03 AAH51112-14 AAH51112-06
3-Iodophenylboronic acid 97%
3-Iodophenylboronic acid

Purity: 97% CAS No.: 221037-98-5 Molecular Formula: C6H6BIO2 Formula Weight: 247.83 MDL No.: MFCD01319015

Formula: C₆H₆BIO₂
MW: 247.83 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01319015
CAS Number: 221037-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR