5-Quinolinol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Hydroxyquinoline, Quinolin-5-ol

H51096-03 H51096-MD
AAH51096-03EA 78.3 USD
AAH51096-03 AAH51096-MD
5-Quinolinol 99%
5-Quinolinol

Purity: 99% CAS No.: 578-67-6 Molecular Formula: C9H7NO Formula Weight: 145.16 MDL No.: MFCD00006792

Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 223 - 226 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006792
CAS Number: 578-67-6
EINECS: 209-428-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR