2-Amino-5-fluorobenzothiazole 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-Fluorobenzo[d]thiazol-2-amine

H51022-03 H51022-MD
AAH51022-03EA 203 USD
AAH51022-03 AAH51022-MD
2-Amino-5-fluorobenzothiazole 98%
2-Amino-5-fluorobenzothiazole

Purity: 98% CAS No.: 20358-07-0 Molecular Formula: C7H5FN2S Formula Weight: 168.19 MDL No.: MFCD05664560

Formula: C₇H₅FN₂S
MW: 168.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05664560
CAS Number: 20358-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR