3-Amino-4-iodopyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Iodo-3-pyridylamine, 4-Iodopyridin-3-amine, 4-Iodo-pyridin-3-ylamine
H50023-03
H50023-06
AAAH50023-03EA
146
USD
AAAH50023-03
AAAH50023-06
3-Amino-4-iodopyridine 98%
3-Amino-4-iodopyridine
Purity: 98%
CAS No.: 105752-11-2
Molecular Formula: C5H5IN2
Formula Weight: 220.01
Melting Point: 55-68°
MDL No.: MFCD04971334
Formula:
C₅H₅IN₂ MW: 220.01 Melting Pt: 55…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD04971334 CAS Number: 105752-11-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...