7-Bromoisatin 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 7-Bromo-1H-indole-2,3-dione, 7-Bromoindoline-2,3-dione

H32843-03 H32843-09
AAH32843-03EA 32.9 USD
AAH32843-03 AAH32843-09
7-Bromoisatin 97%
7-Bromoisatin

Purity: 97% CAS No.: 20780-74-9 Molecular Formula: C8H4BrNO2 Formula Weight: 226.03 Melting Point: 195-199° MDL No.: MFCD00774354

Formula: C₈H₄BrNO₂
MW: 226.03 g/mol
Melting Pt: 195…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00774354
CAS Number: 20780-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR