5-Methylpyridin-3-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Hydroxy-5-methylpyridine, 3-Hydroxy-5-picoline

H32658-06 H32658-03
AAH32658-MDEA 113.06 USD
AAH32658-MD AAH32658-06 AAH32658-03
5-Methylpyridin-3-ol 97%
5-Methylpyridin-3-ol

Purity: 97% CAS No.: 42732-49-0 Molecular Formula: C6H7NO Formula Weight: 109.13 Melting Point: 136-140° MDL No.: MFCD00661297

Formula: C₆H₇NO
MW: 109.13 g/mol
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00661297
CAS Number: 42732-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR