2-Chloro-6-fluorobenzothiazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-6-fluorobenzo[d]thiazole

H32383-06 H32383-03
AAH32383-06EA 265 USD
AAH32383-06 AAH32383-03
2-Chloro-6-fluorobenzothiazole 98%
2-Chloro-6-fluorobenzothiazole

Purity: 98% CAS No.: 399-74-6 Molecular Formula: C7H3ClFNS Formula Weight: 187.62 Melting Point: 89-93° MDL No.: MFCD04448822

Formula: C₇H₃ClFNS
MW: 187.62 g/mol
Melting Pt: 89…93 °C
Storage Temperature: Ambient
MDL Number: MFCD04448822
CAS Number: 399-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR