4-Iodobutyl acetate 96% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Acetic acid 4-iodobutyl ester
Stabilization:
Stabilised with copper
H31826-09
H31826-18
AAH31826-09EA
70.6
USD
AAH31826-09
AAH31826-18
4-Iodobutyl acetate 96% stabilised
4-Iodobutyl acetate
Purity: 96%
CAS No.: 40596-44-9
Molecular Formula: C6H11IO2
Formula Weight: 242.05
Storage Temperature: 2°C to 8°C
Flash Point: 91°(196°F)
Refractive Index: 1.5115
Boiling Point: 94-95°/5.5mm
MDL No.: MFCD00010664
Formula:
C₆H₁₁IO₂ Boiling Pt: 94…95 °C/5.5 mm Density: 1.610 Flash Pt: 91 °C(196 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010664 CAS Number: 40596-44-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...