3-Fluoro-2-hydroxypyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H31705-03
H31705-MD
H31705-06
AAH31705-03EA
144
USD
AAH31705-03
AAH31705-MD
AAH31705-06
3-Fluoro-2-hydroxypyridine 97%
3-Fluoro-2-hydroxypyridine
Purity: 97%
CAS No.: 1547-29-1
Molecular Formula: C5H4FNO
Formula Weight: 113.09
Odor: Odorless
Melting Point: 157-161°
MDL No.: MFCD03095042
Formula:
C₅H₄FNO Melting Pt: 157…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095042 CAS Number: 1547-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...