Methyl oleate 96%

Supplier: Thermo Scientific Chemicals

Synonyms: cis-Oleic Acid Methyl Ester, Oleic acid methyl ester

H31358-14 H31358-22
AAH31358-14EA 286.98 USD
AAH31358-14 AAH31358-22
Methyl oleate 96%
Methyl oleate

Purity: 96% CAS No.: 112-62-9 Molecular Formula: C19H36O2 Formula Weight: 296.50 Flash Point: 163°(325°F) Refractive Index: 1.4520 Boiling Point: 168-170°/2mm Beilstein Reference: 1727037 Merck Reference: 14,6828 MDL No.: MFCD00009578

Formula: C₁₉H₃₆O₂
MW: 296.49 g/mol
Boiling Pt: 218.5
Melting Pt: -20
Density: 0.876
Flash Pt: 163 °C(325 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009578
CAS Number: 112-62-9
EINECS: 203-992-5
Merck Index: 14,6828

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR