4'-Hydroxy-6-methylflavone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Methyl-2-(4-hydroxyphenyl)-4-benzopyrone, 6-Methyl-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 6-Methyl-2-(4-hydroxyphenyl)chromen-4-one

H30959-06 H30959-03 H30959-14
AAH30959-06EA 71.5 USD
AAH30959-06 AAH30959-03 AAH30959-14
4'-Hydroxy-6-methylflavone 97%
4'-Hydroxy-6-methylflavone

Purity: 97% CAS No.: 288401-04-7 Molecular Formula: C16H12O3 Formula Weight: 252.28 Melting Point: 238-243° MDL No.: MFCD03424432

Formula: C₁₆H₁₂O₃
MW: 252.27 g/mol
Melting Pt: 238…243 °C
Storage Temperature: Ambient
MDL Number: MFCD03424432
CAS Number: 288401-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR