4-Ethylbenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (4-Ethylphenyl)methanol

H30459-06 H30459-03
AAH30459-06EA 148 USD
AAH30459-06 AAH30459-03
4-Ethylbenzyl alcohol 99%
4-Ethylbenzyl alcohol

Purity: 99% CAS No.: 768-59-2 Molecular Formula: C9H12O Formula Weight: 136.19 Refractive Index: 1.5275 Boiling Point: 115-117°/9mm MDL No.: MFCD00004666

Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 115…117 °C/9 mm
Density: 1.028
Storage Temperature: Ambient
MDL Number: MFCD00004666
CAS Number: 768-59-2
EINECS: 292-198-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR