(±)-3-Hydroxybutyric acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Hydroxybutyric acid, DL-3-Hydroxybutyric Acid, (±)-3-Hydroxybutanoic acid
H30426-06
H30426-14
AAH30426-06EA
216
USD
AAH30426-06
AAH30426-14
(±)-3-Hydroxybutyric acid 98%
(±)-3-Hydroxybutyric acid
Purity: 98%
CAS No.: 300-85-6
Molecular Formula: C4H8O3
Formula Weight: 104.10
Odor: Odorless
Melting Point: 36-39°
MDL No.: MFCD00004546
Formula:
C₄H₈O₃ MW: 104.11 g/mol Boiling Pt: 269 °C (1013 hPa) Melting Pt: 33 - 34 °C Density: 1.26 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004546 CAS Number: 300-85-6 EINECS: 206-099-9 Merck Index: 12,04861 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...