4-Bromo-2-fluoro-6-nitroanisole 97%

Supplier: Thermo Scientific Chemicals
Warning

H30220-14 H30220-06
AAH30220-14EA 295 USD
AAH30220-14 AAH30220-06
4-Bromo-2-fluoro-6-nitroanisole 97%
4-Bromo-2-fluoro-6-nitroanisole

Purity: 97% CAS No.: 74266-66-3 Molecular Formula: C7H5BrFNO3 Formula Weight: 250.02 Melting Point: 53-56° MDL No.: MFCD00013374

Formula: C₇H₅BrFNO₃
MW: 250.02 g/mol
Melting Pt: 53…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00013374
CAS Number: 74266-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR