2-Amino-3-chloropyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

H30166-03 H30166-06
AAH30166-03EA 134 USD
AAH30166-03 AAH30166-06 AAH30166-MD
2-Amino-3-chloropyridine 95%
2-Amino-3-chloropyridine

Purity: 95% CAS No.: 39620-04-7 Molecular Formula: C5H5ClN2 Formula Weight: 128.56 Melting Point: 59-62° MDL No.: MFCD03541052

Formula: C₅H₅ClN₂
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD03541052
CAS Number: 39620-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR