2-Amino-5-chloro-3-fluoropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H30135-03 H30135-06
AAH30135-03EA 113 USD
AAH30135-03 AAH30135-06 AAH30135-MD
2-Amino-5-chloro-3-fluoropyridine 98%
2-Amino-5-chloro-3-fluoropyridine

Purity: 98% CAS No.: 246847-98-3 Molecular Formula: C5H4ClFN2 Formula Weight: 146.55 Melting Point: 95-97° MDL No.: MFCD09878434

Formula: C₅H₄ClFN₂
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD09878434
CAS Number: 246847-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR