Benzophenone imine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H30110-06
H30110-14
AAH30110-06EA
68.5
USD
AAH30110-06
AAH30110-14
Benzophenone imine 95%
Benzophenone imine
Purity: 95%
CAS No.: 1013-88-3
Molecular Formula: C13H11N
Formula Weight: 181.23
Refractive Index: 1.6180
Boiling Point: 151-153°/10mm
MDL No.: MFCD00001760
Formula:
C₁₃H₁₁N Boiling Pt: 151…153 °C/10 mm Density: 1.08 Storage Temperature: Ambient |
MDL Number:
MFCD00001760 CAS Number: 1013-88-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...