7-Methylxanthine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,6-Dihydroxy-7-methylpurine, 7-Methyl-3,7-dihydro-1H-purine-2,6-dione, 3,7-Dihydro-7-methyl-1H-purine-2,6-dione

H29190-MD H29190-MF
AAH29190-03EA 546.39 USD
AAH29190-03 AAH29190-MD AAH29190-MF
7-Methylxanthine 98%
7-Methylxanthine

Purity: 98% CAS No.: 552-62-5 Molecular Formula: C6H6N4O2 Formula Weight: 166.14 Melting Point: 300° MDL No.: MFCD00037979

Formula: C₆H₆N₄O₂
MW: 166.14 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00037979
CAS Number: 552-62-5
EINECS: 209-019-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR