2-Cyanobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Formylbenzonitrile

H29027-03 H29027-06
AAH29027-03EA 52 USD
AAH29027-03 AAH29027-06
2-Cyanobenzaldehyde 98%
2-Cyanobenzaldehyde

Purity: 98% CAS No.: 7468-67-9 Molecular Formula: C8H5NO Formula Weight: 131.13 Odor: Characteristic Melting Point: 102-106° MDL No.: MFCD00017503

Formula: C₈H₅NO
MW: 131.13 g/mol
Melting Pt: 102…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00017503
CAS Number: 7468-67-9
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR